4-(9-Carbazolyl)benzeneboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(9H-9-carbozale)phenylboronic acid, 4-(9H-Carbazol-9-yl)phenylboronic acid, (4-(9H-Carbazol-9-yl)phenyl)boronic acid, 4-(9H-Carbazol-9-yl)benzeneboronic acid
A151461-100G
A151461-10G
A151461-25G
A151461-1G
A151461-5G
103804-256EA
6
USD
103804-256
103842-408
76973-132
76973-134
77299-874
77317-974
77317-976
4-(9-Carbazolyl)benzeneboronic acid 98%
4-(9-Carbazolyl)benzeneboronic acid
Precaución: for Research Use Only
Formula:
C₁₈H₁₄BNO₂ MW: 287.125 g/mol Boiling Pt: 452.7 °C (760 mm Hg) Melting Pt: 264 °C (760 mm Hg) Flash Pt: 227.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD13176534 CAS Number: 419536-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...