4-(Hydroxymethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, 2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Hydroxymethyl)benzeneboronic acid pinacol ester, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 4-(Hydroxymethyl)phenylboronic acid Pinacol ester

A151856-100G A151856-10G A151856-250MG A151856-1G A151856-5G
103804-310EA 9 USD
103804-310 103842-460 76812-226 76812-228 76812-230 77318-042 77318-044
4-(Hydroxymethyl)phenylboronic acid pinacol ester 98%
4-(Hydroxymethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
MW: 234.10 g/mol
Boiling Pt: 353.3 °C (760 mm Hg)
Flash Pt: 167.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09837617
CAS Number: 302348-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR