4-(Methoxycarbonylmethyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-(2-Methoxy-2-oxoethyl)phenyl)boronic acid
A319642-100MG
A319642-10G
A319642-250MG
A319642-1G
A319642-5G
103825-210EA
74
USD
103825-210
103836-142
77002-826
77002-828
77002-830
77306-202
77306-204
4-(Methoxycarbonylmethyl)phenylboronic acid 97%
4-(Methoxycarbonylmethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁BO₄ MW: 193.9922 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09997796 CAS Number: 454185-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...