4-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-(2,2,2-Trifluoroethoxy)phenyl)boronic acid, 4-(2,2,2-Trifluoroethoxy)benzeneboronic acid
A159019-1G
A159019-250MG
103805-212EA
17
USD
103805-212
103843-424
77319-286
77592-932
4-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₈BF₃O₃ MW: 219.95 g/mol Melting Pt: 191…194 °C Flash Pt: 133.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09027239 CAS Number: 886536-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...