(4-(1-Hydroxyethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A289704-1G A289704-5G A289704-100MG A289704-10G A289704-250MG
103813-842EA 21 USD
103813-842 103834-982 76837-566 76837-568 76988-632 76988-634 76988-636
(4-(1-Hydroxyethyl)phenyl)boronic acid 97%
(4-(1-Hydroxyethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04112543
CAS Number: 518336-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR