(4-((Trimethylsilyl)ethynyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A495797-250MG
A495797-1G
103828-252EA
477
USD
103828-252
76830-112
77684-316
(4-((Trimethylsilyl)ethynyl)phenyl)boronic acid 98%
(4-((Trimethylsilyl)ethynyl)phenyl)boronic acid
Precaución: For research use only.
Formula:
C₁₁H₁₅BO₂Si MW: 218.13 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10697430 CAS Number: 630127-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...