4-(Tetrahydropyran-2-yloxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid

A137437-100G A137437-10G A137437-250MG A137437-1G A137437-5G
103802-456EA 6 USD
103802-456 76996-716 76996-718 76996-720 77315-412 77315-414
4-(Tetrahydropyran-2-yloxy)phenylboronic acid 97%
4-(Tetrahydropyran-2-yloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₄
MW: 222.05 g/mol
Boiling Pt: 411.3±55.0 °C (760 mm Hg)
Melting Pt: 117…124 °C
Flash Pt: 202.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02183525
CAS Number: 182281-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR