4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (4-((2-(Dimethylamino)ethyl)carbamoyl)phenyl)boronic acid hydrochloride, 4-Borono-N-[2-(dimethylamino)ethyl]benzamide hydrochloride

A401900-10G A401900-1G A401900-25G A401900-5G
103827-068EA 35 USD
103827-068 103838-104 77291-150 77307-072 77307-074 77307-076
4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride 98%
4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₁₁H₁₇BN₂O₃·ClH
MW: 272.536 g/mol
Melting Pt: 186…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656014
CAS Number: 850568-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR