3-Vinylphenylboronic acid 95%

Supplier: Ambeed
Warning

A201221-100G A201221-10G A201221-250MG A201221-1G A201221-25G A201221-5G
103817-280EA 36 USD
103817-280 103847-918 77248-850 77248-852 77248-854 77302-738 77302-740 77302-742
3-Vinylphenylboronic acid 95%
3-Vinylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₂
Boiling Pt: 320.2 °C (760 mm Hg)
Melting Pt: 127…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074679
CAS Number: 15016-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR