(3S,4R)-tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A217936-100MG A217936-250MG A217936-1G
103818-672EA 206 USD
103818-672 76873-852 76873-854 77303-468
(3S,4R)-tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate 95%
(3S,4R)-tert-Butyl 4-amino-3-methoxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
Boiling Pt: 306.2±42.0 °C (760 mm Hg)
Flash Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784010
CAS Number: 1171125-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR