(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%

Supplier: Ambeed
Warning

Synonyms: (3R,6R)-3,6-Dimethyl-1,4-dioxane-2,5-dione

A178884-100G A178884-10G A178884-25G A178884-1G
103807-382EA 6 USD
103807-382 103845-722 76690-480 76690-482 77083-166 77301-586
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.125 g/mol
Boiling Pt: 285.5 °C (760 mm Hg)
Flash Pt: 150.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00082566
CAS Number: 13076-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR