(3R,5R)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A135685-100MG A135685-250MG A135685-1G A135685-5G
103802-206EA 246 USD
103802-206 76823-214 76823-216 77298-812 77298-814
(3R,5R)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 97%
(3R,5R)-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3 g/mol
Boiling Pt: 279.7 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08686695
CAS Number: 438049-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR