(3R,5R)-3,5-Dimethylmorpholine hydrochloride 95%

Supplier: Ambeed
Warning

A153632-100MG A153632-250MG A153632-1G
103804-542EA 169 USD
103804-542 77010-918 77010-920 77318-384
(3R,5R)-3,5-Dimethylmorpholine hydrochloride 95%
(3R,5R)-3,5-Dimethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914345
CAS Number: 1542268-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR