D-Gulonic acid-γ-lactone 97%
Supplier: Ambeed
|
Warning
|
A433380-100G
A433380-10G
A433380-1G
A433380-25G
103838-618EA
33
USD
103838-618
76695-086
76695-088
76695-090
77307-268
D-Gulonic acid-γ-lactone 97%
D-Gulonic acid-γ-lactone
Precaución: for Research Use Only
Formula:
C₆H₁₀O₆ Boiling Pt: 467.9 °C (760 mm Hg) Melting Pt: 182…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005391 CAS Number: 6322-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...