(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3R,4S,5R,6R)-3,4,5-tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
A215846-25G
A215846-5G
A215846-100G
103849-226EA
5
USD
103849-226
77087-778
77087-780
77568-086
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol 98%
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol
Precaución: For research use only.
Formula:
C₃₄H₃₆O₆ MW: 540.65 g/mol Boiling Pt: 672.4 °C (760 mm Hg) Melting Pt: 145…149 °C Flash Pt: 360.441 °C Storage Temperature: Freezer |
MDL Number:
MFCD00066004 CAS Number: 4132-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...