(3R,4S)-rel-4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A245494-100MG A245494-250MG
103811-650EA 287 USD
103811-650 77153-180 77153-182
(3R,4S)-rel-4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
(3R,4S)-rel-4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₀BrNO₄
MW: 370.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659267
CAS Number: 1161787-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR