D-Galactosamine pentaacetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2,3,4,6-Penta-O-acetyl-D-galactosamine, (3R,4R,5R,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate
A137694-10G
A137694-1G
A137694-25G
A137694-5G
103802-492EA
10
USD
103802-492
76688-864
77315-468
77315-470
77315-472
D-Galactosamine pentaacetate 98%
D-Galactosamine pentaacetate
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NO₁₀ MW: 389.355 g/mol Boiling Pt: 530.2 °C (760 mm Hg) Melting Pt: 230 °C Flash Pt: 274.433 °C Storage Temperature: Freezer |
MDL Number:
MFCD00050926 CAS Number: 76375-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...