Isomalt 97%

Supplier: Ambeed
Warning

Synonyms: Palatinit, Palatinitol, C-Isomaltidex, 1-O-alpha-D-Glucopyranosyl-D-mannitol, 6-O-alpha-L-allopyranosyl-D-glucitol, (3R,4R,5R)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,4,5-pentaol, 6-O-alpha-D-Glucopyranosyl-D-glucitol mixed with 1-O-alpha-D-glucopyranosyl-D-mannitol

A190324-10G A190324-250MG A190324-25G A190324-1G A190324-5G A190324-100G A190324-500G
103816-284EA 6 USD
103816-284 103846-886 76690-850 76690-852 76690-854 77084-676 77084-678 77600-546 77625-914
Isomalt 97%
Isomalt

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₁₁
MW: 344.31 g/mol
Boiling Pt: 788.5 °C (760 mm Hg)
Flash Pt: 430.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00190708
CAS Number: 64519-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR