(3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)boronic acid 95%

Supplier: Ambeed
Warning

A101101-100MG A101101-250MG A101101-1G
103795-802EA 487 USD
103795-802 103821-492 76821-226 76821-228 77282-660
(3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)boronic acid 95%
(3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₄
MW: 192.96 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16036561
CAS Number: 1246765-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR