3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane 95%

Supplier: Ambeed
Warning

Synonyms: (3-Methyloxetan-3-yl)methyl 4-methylbenzenesulphonate

A200660-10G A200660-250MG A200660-1G A200660-25G A200660-5G
103817-230EA 32 USD
103817-230 103847-862 77274-810 77274-812 77302-702 77302-704 77302-706
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane 95%
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄S
MW: 256.316 g/mol
Boiling Pt: 381.1 °C (760 mm Hg)
Melting Pt: 50…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275394
CAS Number: 99314-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR