(3-Methyloxetan-3-yl)methanamine 97%

Supplier: Ambeed
Danger

A243147-5G A243147-10G A243147-250MG A243147-1G A243147-25G
103811-508EA 31 USD
103811-508 103851-332 77090-528 77288-782 77288-784 77304-266 77304-268
(3-Methyloxetan-3-yl)methanamine 97%
(3-Methyloxetan-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.1469 g/mol
Boiling Pt: 134.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08703640
CAS Number: 153209-97-3
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR