(3-Methyl-5-nitrophenyl)methanol 98%

Supplier: Ambeed
Warning

A232301-100MG A232301-250MG A232301-1G A232301-5G
103810-832EA 293 USD
103810-832 103850-584 76987-426 76987-428 77089-512 77089-514
(3-Methyl-5-nitrophenyl)methanol 98%
(3-Methyl-5-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
Melting Pt: 56.8…57.9 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD18399653
CAS Number: 107757-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR