3-Methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)anisole, (3-Methoxyphenyl)methanol

A115246-10G A115246-25G A115246-500G A115246-5G
103824-386EA 5 USD
103824-386 77108-232 77108-234 77108-236 77311-198
3-Methoxybenzyl alcohol 98%
3-Methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
Boiling Pt: 247.4 °C (760 mm Hg)
Melting Pt: 29…30 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004637
CAS Number: 6971-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR