3-Methoxy-4-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

A280234-5G A280234-10G A280234-250MG A280234-25G
103813-500EA 35 USD
103813-500 103853-444 77093-380 77250-494 77250-496 77250-498
3-Methoxy-4-nitrobenzyl alcohol 98%
3-Methoxy-4-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₄
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007098
CAS Number: 80866-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR