3-Methoxy-2-methylbenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Methoxy-2-methylphenyl)boronic acid, 3-Borono-2-methylanisole, 3-Methoxy-2-methylphenylboronic acid, 2-Borono-6-methoxytoluene

A100219-100MG A100219-250MG A100219-5G
103795-304EA 87 USD
103795-304 77148-154 77148-156 77610-728
3-Methoxy-2-methylbenzeneboronic acid 97%
3-Methoxy-2-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
Boiling Pt: 325.8±52.0 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18398348
CAS Number: 1313617-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR