3-Iodophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(3-iodophenyl)borane, (3-Iodophenyl)boronic acid, 3-Iodobenzeneboronic acid, (3-Iodophenyl)metaboric acid

A227422-1G A227422-25G A227422-5G A227422-100G A227422-10G A227422-250MG
103819-318EA 12 USD
103819-318 103850-224 77088-990 77088-992 77088-994 77275-660 77275-662 77275-664
3-Iodophenylboronic acid 98%
3-Iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BIO₂
Boiling Pt: 358.8 °C (760 mm Hg)
Melting Pt: 195…197 °C
Flash Pt: 170.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01319015
CAS Number: 221037-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR