(3-Fluoropyridin-4-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(hydroxymethyl)pyridine

A175623-10G A175623-1G A175623-25G
103807-070EA 10 USD
103807-070 103845-382 76872-574 77082-724 77082-726
(3-Fluoropyridin-4-yl)methanol 97%
(3-Fluoropyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.12 g/mol
Flash Pt: 87.439 °C
Storage Temperature: Ambient
MDL Number: MFCD11656367
CAS Number: 870063-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR