3-Fluorobenzylamine 95%

Supplier: Ambeed
Danger

A114357-25G A114357-1G A114357-5G A114357-10G
103798-390EA 6 USD
103798-390 103824-230 77108-202 77297-304 77311-040 77570-800
3-Fluorobenzylamine 95%
3-Fluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈FN
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008113
CAS Number: 100-82-3
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR