3-Fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

A180833-500G A180833-1G A180833-5G A180833-10G A180833-100G
103807-566EA 5 USD
103807-566 103845-928 77274-068 77301-696 77301-698 77550-166 77748-183
3-Fluorophenylboronic acid 98%
3-Fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Melting Pt: 214…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00236042
CAS Number: 768-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR