(3-Fluorooxetan-3-yl)methyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

A395671-100MG A395671-250MG
103826-964EA 66 USD
103826-964 103838-002 77168-258 77168-260
(3-Fluorooxetan-3-yl)methyl 4-methylbenzenesulfonate 97%
(3-Fluorooxetan-3-yl)methyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄S
MW: 260.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706189
CAS Number: 1308644-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR