3-Fluoro-5-hydroxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-hydroxybenzeneboronic acid, 3-Fluoro-5-hydroxyphenylboronic acid, (3-Fluoro-5-hydroxyphenyl)boronic acid

A157116-100MG A157116-250MG A157116-1G
103804-966EA 28 USD
103804-966 103843-166 76783-656 76783-658 77318-982
3-Fluoro-5-hydroxybenzeneboronic acid 98%
3-Fluoro-5-hydroxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.92 g/mol
Boiling Pt: 380 °C (760 mm Hg)
Melting Pt: 54…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363778
CAS Number: 871329-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR