3-Fluoro-4-methoxybenzylamine 97%

Supplier: Ambeed
Danger

A220615-1G A220615-25G A220615-5G A220615-10G
103818-864EA 14 USD
103818-864 103849-746 77088-336 77088-338 77088-340 77275-480 77275-482
3-Fluoro-4-methoxybenzylamine 97%
3-Fluoro-4-methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
Boiling Pt: 234.9 °C (760 mm Hg)
Flash Pt: 95.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04116360
CAS Number: 123652-95-9
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR