(3-Fluoro-4-hydroxyphenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A146892-10G
A146892-250MG
A146892-1G
A146892-25G
A146892-5G
103803-716EA
15
USD
103803-716
103841-810
76783-204
76783-206
77317-164
77317-166
77317-168
(3-Fluoro-4-hydroxyphenyl)boronic acid 98%
(3-Fluoro-4-hydroxyphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₅H₅BFNO₂ Boiling Pt: 331.5 °C (760 mm Hg) Melting Pt: 252…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659838 CAS Number: 182344-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...