3-Fluoro-4-(trifluoromethyl)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-(trifluoromethyl)phenyl)methanol

A151891-10G A151891-1G A151891-5G
103804-314EA 12 USD
103804-314 103842-464 76997-374 77318-048 77318-050
3-Fluoro-4-(trifluoromethyl)benzyl alcohol 98%
3-Fluoro-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MW: 194.13 g/mol
Boiling Pt: 208.5 °C (760 mm Hg)
Melting Pt: 28…32 °C
Flash Pt: 79.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00236285
CAS Number: 230295-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR