3-Fluoro-2-formylphenylboronic acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid
A137351-10G
A137351-1G
A137351-5G
A137351-100G
103802-446EA
9
USD
103802-446
76811-578
77315-400
77315-402
77611-244
3-Fluoro-2-formylphenylboronic acid 98%
3-Fluoro-2-formylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₆BFO₃ Melting Pt: 123…127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10697421 CAS Number: 871126-15-7 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...