3-Ethyl-3-oxetanemethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane
A136980-100G
A136980-10G
A136980-1G
A136980-500G
A136980-25G
A136980-5G
103808-548EA
79
USD
103808-548
77272-304
77272-306
77272-308
77272-310
77298-876
77298-878
3-Ethyl-3-oxetanemethanol 98%
3-Ethyl-3-oxetanemethanol
Precaución: for Research Use Only
Formula:
C₆H₁₂O₂ Boiling Pt: 203 °C (760 mm Hg) Flash Pt: 86.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00190143 CAS Number: 3047-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...