(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: (3-Ethoxy-3-oxopropyl)triphenylphosphonium bromide

A163036-100G A163036-10G A163036-250MG A163036-1G A163036-25G A163036-5G
103805-680EA 6 USD
103805-680 103843-938 77247-704 77247-706 77247-708 77320-016 77320-018 77320-020
(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide 97%
(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide

Precaución: for Research Use Only

Formula: C₂₃H₂₄O₂P·Br
MW: 443.313 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701355
CAS Number: 42843-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR