3-Cyano-4-pyridineboronic acid 95+%
Supplier: Ambeed
|
Warning
|
A151096-250MG
A151096-5G
A151096-100MG
103804-230EA
483
USD
103804-230
77871-744
77871-745
77871-743
3-Cyano-4-pyridineboronic acid 95+%
3-Cyano-4-pyridineboronic acid
Precaución: Inert atmosphere, 2…8 °C.
Formula:
C₆H₅BN₂O₂ Boiling Pt: 407.9 °C (760 mm Hg) Flash Pt: 200.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09037480 CAS Number: 874290-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...