3-Cyano-5-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Cyano-5-fluorophenyl)boronic acid

A197102-10G A197102-250MG A197102-1G A197102-25G A197102-5G
103816-890EA 25 USD
103816-890 103847-506 76691-078 76691-080 77085-488 77085-490 77085-492
3-Cyano-5-fluorophenylboronic acid 97%
3-Cyano-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MW: 164.9300 g/mol
Boiling Pt: 343.1 °C (760 mm Hg)
Melting Pt: >320 °C
Flash Pt: 161.335 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263608
CAS Number: 304858-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR