3-Cyano-2-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Cyano-2-fluorophenyl)boronic acid

A418505-250MG A418505-1G A418505-5G
103827-356EA 33 USD
103827-356 103838-400 76829-426 76842-004 76842-006
3-Cyano-2-fluorophenylboronic acid 98%
3-Cyano-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MW: 164.9295 g/mol
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD07374984
CAS Number: 957121-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR