(3-Chloropyrazin-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A239564-100G A239564-10G A239564-250MG A239564-500G A239564-1G A239564-25G A239564-5G
103811-298EA 6 USD
103811-298 103851-096 76976-410 76976-412 76976-414 76976-416 77090-236 77090-238 77090-240
(3-Chloropyrazin-2-yl)methanamine hydrochloride 95%
(3-Chloropyrazin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇Cl₂N₃
MW: 180.0352 g/mol
Melting Pt: 222…226 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09910171
CAS Number: 939412-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR