(3-Chloropyrazin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-3-chloropyrazine dihydrochloride

A101257-10G A101257-250MG A101257-1G A101257-5G A101257-25G
103795-872EA 23 USD
103795-872 103821-556 76970-146 76970-148 77282-688 77282-690 77670-462
(3-Chloropyrazin-2-yl)methanamine dihydrochloride 95%
(3-Chloropyrazin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₈Cl₃N₃
MW: 216.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07644629
CAS Number: 867165-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR