(3-Chlorobenzyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A191244-100MG A191244-10G A191244-250MG A191244-1G
103816-356EA 33 USD
103816-356 103846-958 77164-280 77164-282 77164-284 77302-250
(3-Chlorobenzyl)hydrazine hydrochloride 95%
(3-Chlorobenzyl)hydrazine hydrochloride

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₇H₁₀Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD13186199
CAS Number: 260057-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR