3-Chloro-5-hydroxyphenylboronic acid 95%

Supplier: Ambeed
Warning

A107342-100MG A107342-250MG A107342-1G A107342-5G
103797-172EA 131 USD
103797-172 103822-940 76970-482 76970-484 77309-786 77309-788
3-Chloro-5-hydroxyphenylboronic acid 95%
3-Chloro-5-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
Boiling Pt: 403.9±55.0 °C (760 mm Hg)
Flash Pt: 198.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15143456
CAS Number: 1214900-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR