3-Chloro-5-(trifluoromethyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-Chloro-5-(trifluoromethyl)phenyl)boronic acid
A395852-25G
A395852-5G
A395852-10G
A395852-250MG
103826-968EA
14
USD
103826-968
103838-006
76841-426
76841-428
77278-702
77278-704
3-Chloro-5-(trifluoromethyl)phenylboronic acid 97%
3-Chloro-5-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BClF₃O₂ MW: 224.3700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07778020 CAS Number: 1160561-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...