(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A873244-1G A873244-5G A873244-10G A873244-25G
103841-290EA 50 USD
103841-290 76845-548 76845-550 76981-854 76981-856
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid

Precaución: For research use only

Formula: C₈H₉BClFO₃
MW: 218.4177 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15144702
CAS Number: 909122-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR