(3-Carbamoylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-carbamoylphenyl)boronic acid, 3-Aminocarbonylphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-Carbamoylphenylboronic acid, 3-Carbamoylbenzeneboronic acid

A229957-1G A229957-10G A229957-25G
103819-510EA 5 USD
103819-510 103850-412 77089-266 77593-490 77615-912
(3-Carbamoylphenyl)boronic acid 97%
(3-Carbamoylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₃
Melting Pt: 232…235 °C
Flash Pt: 200.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411948
CAS Number: 351422-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR