3-Bromo-1-(trimethylsilyl)-1-propyne 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane

A116676-10G A116676-25G A116676-5G A116676-100G
103798-782EA 5 USD
103798-782 103824-648 77271-234 77271-236 77311-440 77571-008
3-Bromo-1-(trimethylsilyl)-1-propyne 97%
3-Bromo-1-(trimethylsilyl)-1-propyne

Precaución: for Research Use Only

Formula: C₆H₁₁BrSi
Boiling Pt: 195.8 °C (760 mm Hg)
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134460
CAS Number: 38002-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR