3-Bromobenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (3-Bromophenyl)methanamine

A519645-25G A519645-5G A519645-10G A519645-1G
103828-494EA 9 USD
103828-494 103839-602 76843-604 76843-606 77116-682 77307-710
3-Bromobenzylamine 98%
3-Bromobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 244.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01026119
CAS Number: 10269-01-9
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR