(3-Bromo-4-methylphenoxy)(tert-butyl)dimethylsilane 97%

Supplier: Ambeed
Warning

A271820-10G A271820-1G
103813-068EA 6 USD
103813-068 103852-980 76988-322 77092-768
(3-Bromo-4-methylphenoxy)(tert-butyl)dimethylsilane 97%
(3-Bromo-4-methylphenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₁BrOSi
MW: 301.3 g/mol
Boiling Pt: 293.6±28.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD24847837
CAS Number: 164513-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR